Learn More
Thermo Scientific™ CCDA Selective Medium
Isolate Campylobacter spp. with a blood free selective medium which conforms to ISO 10272-1 and 10272-2.
Brand:  Thermo Scientific™ PO5091A
10.84 EUR valid until 2024-12-31
Use promo code "21974" to get your promotional price.
 View more versions of this product
Product Code. 12815093
  • 12.55€ / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Isolate Campylobacter spp. using blood-free Thermo Scientific™ CCDA Selective Medium, which conforms to ISO 10272-1 and 10272-2.
Modified CCDA Medium is based on the original formulation described by Bolton which was developed to replace blood with charcoal, ferrous sulphate and sodium pyruvate. Improved selectivity is achieved using cefoperazone as the selective agent. Amphotericin B has been added to the formula to suppress the growth of yeast and fungal contaminants that may occur at 37°C.
Campylobacter infections are among the most common bacterial infections, producing both gastrointestinal and systemic illnesses. Campylobacter jejuni is the most commonly reported bacterial cause of foodborne infection in the United States. Campylobacter are small, non-sporeforming, helical, curved gram-negative rods that require a microaerophilic environment and cannot tolerate drying.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
According ISO 10271-a and ISO 10272-2
2° to 12°C, in the dark
Germany
Agar
Flow Wrapped - Single Layer
Jet black, opaque
90mm Monoplate
CCDA Selective Medium
17 g ± 5 %
10/Pk.