missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBR4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13725-1-AP-150UL
This item is not returnable.
View return policy
Description
The heteroteramer with HSD17B8 has NADH-dependent 3-ketoacyl-acyl carrier protein reductase activity. May play a role in biosynthesis of fatty acids in mitochondria. The homotetramer may act as NADPH-dependent quinone reductase. Has broad substrate specificity and reduces 9,10-phethrenequinone, 1,4-benzoquinone and various other o-quinones and p-quinones (in vitro).Specifications
| CBR4 | |
| Polyclonal | |
| Unconjugated | |
| CBR4 | |
| carbonyl reductase 4, CBR4, Quinone reductase CBR4, SDR45C1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 234309, 359725, 84869 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7TS56, Q8N4T8, Q91VT4 | |
| Cbr4 | |
| CBR4 Fusion Protein Ag4678 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction