Learn More
Karl Hecht™ Cavity Slide/Plates
Perfectly suited for holding liquids and cultures for microscopic examination. Karl Hecht™ Cavity Slides/Plates are available with one or more cavities and in single slide or plate format.
Brand:  Karl Hecht™ 42418010
Product Code. 11740555
Description
Description
  • Microscope slides/plates with one or more cavities.
  • Ground edges.
Specifications
Specifications
140 mm
Yes
Microscope Slide
16mm cavity diameter, 1.6mm cavity depth, polished cavities
85 mm
3 mm
24
140 x 85 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Karl Hecht™ Cavity Slide/Plates > Description: 16mm cavity diameter, 1.6mm cavity depth, polished cavities, Dimensions: 140mm x 85mm, Surface Treatment: Frosted, Thickness: 3mm, Well Count: 24