Learn More
Fisherbrand™ Catalyst Platinum-Cured Silicone Formulation Stopped Tubing Assembly
Preconfigured ready-to-use links and assemblies.
Brand:  Fisherbrand™ 97001-77
 View more versions of this product
Product Code. 15571355
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
Preconfigured ready-to-use links and assemblies.
  • Available in Platinum-Cured Silicone, TPEs, and PVC
  • Ensures optimal amount of tubing is in pump head
Specifications
Specifications
Stopped Tubing Assembly
6.4 mm
2.4 mm