Learn More
Löser™ Freezing Needle for Osmometer
Provides initial freezing point trigger for osmometers. Freezing Needling for Osmometer can be used with all osmometers with a sample volume between 50 and 200μL.
Brand: Löser™ 1.05.0400
Description
- Needle for freezing initiation of all Osmometers with sample volume 50 to 200μL.
Specifications
Freezing Needle | |
All Osmometers 50 to 200 μL Sample Volume | |
Triggering the freezing with freezing point Osmometers |