Learn More
Veolia Water Systems™ Cartidges
Robust and reliable water purification cartridges ideal for removing ionic contaminants to achieve specified water quality.
Brand:  Veolia Water Systems™ LC208
 View more versions of this product
Product Code. 12003283
  • 574.00€ / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Water purification cartridge
  • Removes ionic contaminants to achieve specified water quality
  • Compatible with PURELAB Flex
Specifications
Specifications
LC208