Learn More
Carl Zeiss™ Reflector Module
FL, EP and P & C
Brand:  Carl Zeiss™ 424931-0000-000
Product Code. 11822402
  • 312.00€ / Each
Estimated Shipment: 10-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Reflection avoided through tilted mount for emission filters
  • Maximum filter thickness: 5mm
Specifications
Specifications
Gooseneck Light Guide, Reflector Module
0.5 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Carl Zeiss™ Reflector Module > Goose-neck light guide, two branches