Learn More
Carl Zeiss™ Microscope Eyepiece Reticles
Designed for use with Stemi Microscopes
Brand:  Carl Zeiss™ 474068-0000-000
 View more versions of this product
Product Code. 11872092
  • 314.00€ / Each
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Microscope Eyepiece Reticles
  • For use with Stemi™ 1000, 2000 and 2000C
Specifications
Specifications
Net Micrometer
0.74 in.
26 mm
Stemi™ 1000, 2000 and 2000C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products