missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Cariporide
GREENER_CHOICE

Product Code. p-5546143
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18726228

Brand: Tocris Bioscience™ 5358/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective NHE1 inhibitor; cardioprotective and antitumor

Cariporide is a selective Na+/H+ exchanger isoform 1 (NHE1) inhibitor (IC50 values are 0.05, 3 and 1000 μM for NHE1, NHE3 and NHE2 respectively). Attenuates ischemia-induced cardiomyocyte apoptosis in vitro. Reduces cardiac arrhythmia in vivo. Also promotes apoptosis in cancer cells overexpressing NHE1. Orally active.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type Cariporide
Purity >99%
Molecular Weight (g/mol) 283.35
Inhibitors Cariporide

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.