Learn More
Carbonic Anhydrase XIII Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA580389
Additional Details : Weight : 0.01000kg
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Carbonic Anhydrase XIII/CA13.
Carbonic anhydrases (CAs) are a family of zinc metalloenzymes. For background information on the CA family, see MIM 114800.[supplied by OMIM, Mar 2008].Specifications
Carbonic Anhydrase XIII | |
Polyclonal | |
Unconjugated | |
Ca13 | |
2310075C21Rik; Ca13; Car13; Carbonate dehydratase XIII; carbonic anhydrase 13; Carbonic anhydrase XIII; CAXIII; CA-XIII; RGD1560453 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
377677 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with no preservative | |
Q8N1Q1 | |
Ca13 | |
Recombinant Human Carbonic Anhydrase XIII/CA13 protein (Met1-His262) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |