Learn More
Swann-Morton™ Carbon Steel Foil Packets of 5, Non-sterile Scalpel Blades, No. 15C
Longer, more extended cutting edge than the traditional No. 15 blade.
Brand:  Swann-Morton™ 0121
Product Code. 15706799
  • 24.80€ / Pack of 100
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Description
  • Provides additional reach, especially when combined with the ergonomically designed B3 handle.
  • Longer, more extended cutting edge than the traditional No. 15 blade.
  • Used by dentists carrying out periodontal procedures.
  • Used by podiatrists for nucleating corns around the webbing of the toes.
  • Fitment No. 3.
  • Fits handles 3, 3L, 3 graduated, 5B, 7, 9, and B3.
Specifications
Specifications
Carbon Steel
Non-sterile
5/Pk., 100/Box