Learn More
Labconco™ Carbon Pre-Filter
This Carbon Pre-Filter is a spare part for WaterPro RO Stations.
Brand:  Labconco™ 9059400
170.61 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.80000kg
Product Code. 11854750
  • 202.00€ / Each
Estimated Shipment: 18-09-2024
to see stock.
Description
Description
  • Spare Part
  • Required for operation of WaterPro RO Stations
Specifications
Specifications
WaterPro RO Stations
20 in.