missing translation for 'onlineSavingsMsg'
Learn More

Carbon, Activated, Powder, J.T. Baker™

Product Code. 10519190
Click to view available options
Quantity:
500 g
Packaging:
Wide Mouth Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.011
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Packaging Wide Mouth Amber Glass Bottle
Quantity 500 g
Formula Weight 12.01
Physical Form Powder
Chemical Name or Material Carbon, Activated
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'