missing translation for 'onlineSavingsMsg'
Learn More

Captan-d6, Honeywell Fluka™

Product Code. 15960252
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Analytical standard, reference material

Ensure you have the information you need to quickly start your analysis, and pass relevant audits:

  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY

Specifications

CAS 1330190-00-5
Packaging Glass Bottle
Quantity 10 mg
Grade Analytical Standard
For Use With (Application) Fungicides and Bactericides
Physical Form Solid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'