Learn More
Erlab™ Captair™ Work Surfaces
Designed for laboratories specializing in fields such as biology, botany, aerospace, electronics, pharmaceutical and cosmetics. Erlab™ Captair™ Work Surfaces are for use with Captair PCR work stations.
Brand: Erlab™ I0120020002
This item is not returnable.
View return policy
Description
- High chemical and mechanical resistance
- Rounded corners to facilitate cleaning operations
- Built in spill tray
Specifications
For Captair™ Flex™ Model XLS 714 fume hood | |
No. 1 Sealed Tempered Glass Retention Tray, Embedded Metal, Sealed Frame | |
Work surface No. 1 | |
Glass |