missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Cap for Wash Bottle
Click to view available options
Closure Color:
Green
For Use With (Equipment):
53mm Diameter Wash Bottles
Description
- Replacement Parts for multiple families of wash bottles.
Specifications
Specifications
Color | Green |
No. per Pack | 1Pack |
Quantity | 1Pack |
Closure Color | Green |
Product Type | Cap |
For Use With (Equipment) | 53mm Diameter Wash Bottles |
Closure Type | Cap |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'