Learn More
SI Analytics™ Cannon-Fenske Reverse Flow Viscometers
Reverse flow viscometers are calibrated for manual measurement
Brand:  SI Analytics™ 285403018
Product Code. 10263881
  • 408.00€ / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Calibrated for manual measurement
  • Cannon-Fenske reverse flow
  • Type No. 511 03
  • Capillary No. 50
  • Constant K = 0.004
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cannon-Fenske Reverse Flow Viscometer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products