missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cannabinoid R2/CB2/CNR2 Antibody (352110), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB36551MFV450
This item is not returnable.
View return policy
Description
Cannabinoid R2/CB2/CNR2 Monoclonal antibody specifically detects Cannabinoid R2/CB2/CNR2 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
Cannabinoid R2/CB2/CNR2 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
0.1 mL | |
GPCR, Neuroscience, Vision | |
1269 | |
Store at 4C in the dark. |
Flow Cytometry, CyTOF | |
352110 | |
Flow Cytometry, CyTOF-ready | |
cannabinoid receptor 2 (macrophage), CB-2, CB2cannabinoid receptor 2, CX5testis-dominant CNR2 isoform CB2, hCB2 | |
Human embryonic kidney cell line HEK293-derived transfected with human Cannabinoid R2/CB2/CNR2, Met1-Cys360, Accession # NP_001832 | |
RUO | |
Primary | |
Human | |
IgG2b |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cannabinoid R2/CB2/CNR2 Antibody (352110), mFluor Violet 450 SE, Novus Biologicals™ > 0.1 mL; mFluor Violet 450 SE
Spot an opportunity for improvement?Share a Content Correction