missing translation for 'onlineSavingsMsg'
Learn More

Cannabinoid R2/CB2/CNR2 Antibody (352110), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals FAB36551E

Product Code. 18552719

  • 623.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Cannabinoid R2/CB2/CNR2 Monoclonal antibody specifically detects Cannabinoid R2/CB2/CNR2 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cannabinoid R2/CB2/CNR2
Monoclonal
DyLight 405
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry, CyTOF
352110
Flow Cytometry, CyTOF-ready
cannabinoid receptor 2 (macrophage), CB-2, CB2cannabinoid receptor 2, CX5testis-dominant CNR2 isoform CB2, hCB2
Human embryonic kidney cell line HEK293-derived transfected with human Cannabinoid R2/CB2/CNR2, Met1-Cys360, Accession # NP_001832
0.1 mL
GPCR, Neuroscience, Vision
1269
Store at 4°C in the dark.
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cannabinoid R2/CB2/CNR2 Antibody (352110), DyLight 405, Novus Biologicals™ > 0.1 mL; DyLight 405

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.