missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cannabinoid R1/CB1/CNR1 Antibody (368302), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3834I
This item is not returnable.
View return policy
Description
Cannabinoid R1/CB1/CNR1 Monoclonal antibody specifically detects Cannabinoid R1/CB1/CNR1 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
Cannabinoid R1/CB1/CNR1 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
368302 | |
Flow Cytometry, CyTOF-ready | |
CANN6CB1R, cannabinoid receptor 1 (brain), CB1A, CB1K5, CB-RCB1cannabinoid receptor 1, central cannabinoid receptor, CNR | |
HEK293 human embryonic kidney cell line transfected with human Cannabinoid R1/CB1/CNR1, Met1-Leu472, Accession # NP_057167 | |
0.1 mL | |
Cancer, GPCR, Neuroscience, Neurotransmission, Vision | |
1268 | |
Store at 4°C in the dark. | |
IgG2a |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cannabinoid R1/CB1/CNR1 Antibody (368302), Janelia Fluor™ 549, Novus Biologicals™ > 0.1 mL; Janelia Fluor 549
Spot an opportunity for improvement?Share a Content Correction