missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cannabinoid R1/CB1/CNR1 Antibody (368302), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3834W
This item is not returnable.
View return policy
Description
Cannabinoid R1/CB1/CNR1 Monoclonal antibody specifically detects Cannabinoid R1/CB1/CNR1 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
Cannabinoid R1/CB1/CNR1 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
368302 | |
Flow Cytometry, CyTOF-ready | |
CANN6CB1R, cannabinoid receptor 1 (brain), CB1A, CB1K5, CB-RCB1cannabinoid receptor 1, central cannabinoid receptor, CNR | |
HEK293 human embryonic kidney cell line transfected with human Cannabinoid R1/CB1/CNR1, Met1-Leu472, Accession # NP_057167 | |
0.1 mL | |
Cancer, GPCR, Neuroscience, Neurotransmission, Vision | |
1268 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cannabinoid R1/CB1/CNR1 Antibody (368302), DyLight 650, Novus Biologicals™ > 0.1 mL; DyLight 650
Spot an opportunity for improvement?Share a Content Correction