Learn More
Cannabinoid R1/CB1/CNR1 Antibody (368302), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB3834X
Product Code. 18508437
  • 665.00€ / 0.10mL
Explore more special offers
Description
Description
Cannabinoid R1/CB1/CNR1 Monoclonal antibody specifically detects Cannabinoid R1/CB1/CNR1 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cannabinoid R1/CB1/CNR1
Monoclonal
Alexa Fluor 532
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry, CyTOF
368302
Flow Cytometry, CyTOF-ready
CANN6CB1R, cannabinoid receptor 1 (brain), CB1A, CB1K5, CB-RCB1cannabinoid receptor 1, central cannabinoid receptor, CNR
HEK293 human embryonic kidney cell line transfected with human Cannabinoid R1/CB1/CNR1, Met1-Leu472, Accession # NP_057167
0.1 mL
Cancer, GPCR, Neuroscience, Neurotransmission, Vision
1268
Store at 4°C in the dark.
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Cannabinoid R1/CB1/CNR1 Antibody (368302), Alexa Fluor™ 532, Novus Biologicals™ > 0.1 mL; Alexa Fluor 532