missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals cAMP Dependent Protein Kinase Inhibitor (PKI 5-24)

For cellular and molecular biology applications

110.00€ - 129.00€

Specifications

Purification Method HPLC
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 2222.40
Product Type cAMP Dependent Protein Kinase Inhibitor (PKI 5-24)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15440587
View Documents
Thermo Scientific Chemicals
J66438.|D
0.5 mg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
15450587
View Documents
Thermo Scientific Chemicals
1 mg
129.00€
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

cAMP Dependent Protein Kinase Inhibitor (PKI 5-24) acts as inhibitor of c-AMP-dependent protein kinase These proteins bind to the catalytic subunit of cyclic-AMP dependent protein kinase, displacing the regulatory subunit and inhibiting phosphorylating activity. The cyclic-AMP binding capacity of the regulatory subunit is enhanced.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

HPLC
Solid
cAMP Dependent Protein Kinase Inhibitor (PKI 5-24)
Thr-Thr-Tyr-Ala-Asp-Phe-Ile-Ala-Ser-Gly-Arg-Thr-Gly-Arg-Arg-Asn-Ala-Ile-His-Asp
-30°C to -10°C
2222.40
RUO - research use only
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only