Learn More
Unity Lab Services™ Calibration Report Bath
ULS offers a full range of support services and programs for all instruments and supplies, regardless of manufacturer. Maximize your technology investments by anticipating and avoiding problems before they occur.
Brand: Unity Lab Services™ UMV-P9312
This item is not returnable.
View return policy
Description
- Stability control at a temperature setpoint in the center of the bath
- Setpoint in the range -35°C to +200°C
- Fluids used: pure alcohol from -35°C to +5°C, water up to +90°C, and silicone oil up to +200°C
- Reference instrument certified by a COFRAC accredited laboratory
Specifications
Thermostatted Baths | |
Calibration Report |