Learn More
Novex™ Calyculin A
Product Code. 10079362
Click to view available options
:
25μg
Product Code. 10079362
Brand: Novex™ PHZ1044
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Calculin A
Calyculin A is a structurally unique marine toxin with potent and specific protein phosphatase inhibiting activity. This compound inhibits PP2A with similar potency to okadaic acid, but inhibits PP1 with a 10- to 100-fold greater potency.
Format/Formulation: Lyophilized
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store in freezer (-5 to -30°C).
Molecular Weight (g/mol) 1009.2 Da
Gene ID (Entrez) 5519
Shipping Condition Dry Ice
Purification Method TLC
Sterility Non-sterile
Specific Inhibit or Target PP2A and PP1 Phosphatases
Inhibitor Properties Chemical
Inhibitors Phosphatase Inhibitor
Form Liquid
Quantity 25 μg