missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Calcium Test Kits
Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits
174.00€ - 237.00€
Specifications
For Use With (Application) | For waste water, drinking water |
---|---|
Includes | Certificates |
Sufficient For | 100 Tests |
Test Method | Photometric |
Product Code | Brand | Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Range | Price | Quantity & Availability | |||||
10134211
|
WTW™
250428 |
5 to 160 mg/L |
237.00€
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11863921
|
WTW™
252047 |
10 to 250 mg/L |
174.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifications
For waste water, drinking water | |
Certificates | |
100 Tests | |
Photometric |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Calcium Test Method: photometric 5 - 160 mg/l Ca
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : diethylene glycol