missing translation for 'onlineSavingsMsg'
Learn More

Calcium Sulfite, MP Biomedicals

Product Code. 11427914
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 10257-55-3
Quantity 100 g
Molecular Formula CaH2O3S
MDL Number MFCD00040663
Synonym calcium sulfite, calcium sulphite, sulfurous acid, calcium salt 1:1, calciumsulfit, calcium 2+ sulfite
InChI Key GBAOBIBJACZTNA-UHFFFAOYSA-N
SMILES [Ca++].OS(O)=O
IUPAC Name calcium sulfurous acid
Molecular Weight (g/mol) 122.15
PubChem CID 10154041
Formula Weight 120.14
Chemical Name or Material Calcium Sulfite
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'