missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Calcein Violet 450 AM Viability Dye

Product Code. 15570597
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 15570597

Brand: Invitrogen™ 65085439

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Membrane-permeable live-cell labeling dye. Upon entering the cell, intracellular esterases cleave the acetoxymethyl (AM) ester group, yielding the membrane-impermeable Calcein Violet fluorescent dye. Apoptotic and dead cells with compromised cell membranes do not retain Calcein Violet 450.

  • Molecular weight: 600g/mol
  • Peak excitation: 408nm
  • Peak emission: 450nm
  • Reconstitute in high-quality, freshly opened DMSO
  • Once reconstituted, store at −20°C with dessicant and use within a short period of time. Avoid freeze-thawing
  • For fluorescent microscopy, detect using the appropriate filter sets
  • For flow cytometric analysis, excite with the violet laser line (405nm) and detect using filters for Pacific Blue™/eFluor™ 450 (450/50)
  • Co-staining with Annexin V or 7-AAD is recommended to allow the greatest resolution between live and dead/apoptotic cells
TRUSTED_SUSTAINABILITY

Specifications

Color Violet
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Product Type Dye
Emission 408/450
Product Line eBioscience
Quantity 1 mg

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.