Learn More
c-Myb Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17800-1-AP-20UL
 View more versions of this product
Product Code. 16837784
  • 145.00€ / 20µL
Description
Transcriptional activator; DNA-binding protein that specifically recognize the sequence 5′-YAAC[GT]G-3′. Plays an important role in the control of proliferation and differentiation of hematopoietic progenitor cells.
Specifications
Specifications
c-Myb
Polyclonal
Unconjugated
MYB
c myb_CDS, Cmyb, c-Myb, efg, MYB, Proto oncogene c Myb, Transcriptional activator Myb
Rabbit
Antigen Affinity Chromatography
RUO
17863, 4602
-20°C
Liquid
Western Blot, Immunoprecipitation
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P06876, P10242
Myb
c-Myb Fusion Protein Ag12021
20 μL
Primary
Mouse, Rat, Human