Learn More
Wheaton™ C Model Bottom Drive Modular Spacing Roller Culture Apparatus
Product Code. 13433896 Shop All Wheaton Science Products Products
Click to view available options
Includes:
1 Deck
5 Decks
9 Decks
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 13433896
Brand: Wheaton™ WRBMR5090C
Only null left
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
Specifications
Speed Range 0.25 to 8.1 rpm
Electrical Requirements 100/240 V, 50/60 Hz, 35 W
Voltage 100/240 V
Temperature Range (Metric) 10°C to 40°C
Wattage 35 W
Certifications/Compliance CE, CSA, UL
Frequency 50/60 Hz
Depth (Metric) 63 cm
Height (Metric) 179 cm
Width (Metric) 76 cm
Model C Model
For Use With (Application) For laboratory, pilot plant and production cell culture installations
Humidity Range 80% up to 31°C
Includes 9 Decks
Packaging Quantity 1 Ea.
Spacing 17.7 cm