Learn More
Aalborg™ Buna N O-ring Aluminum Frame with Plug
Aalborg™ Buna N O-ring Aluminum Frame with Plug is for use with multiple tube flow meters, which retain most of the unique features associated with single tube units.
Brand: Aalborg™ P31A2-BA2
This item is not returnable.
View return policy
Description
- Rib-guided or fluted metering tubes facilitate stable, accurate readings
- Interchangeability of flow tubes and floats
- Manifolding at inlet or outlet
- Ease of installation and exchange of flow tubes
- Chemical compatibility
- Simple means of mounting
Specifications
3-Tube 150 mm Manifold Mounting Gas/Liquid Flowmeter | |
Frame | |
Plug and Manifold |