missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution, Phthalate Potassium Hydrogen, pH 4.008, Fisher chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

42.00€ - 61.00€

Specifications

Chemical Name or Material Buffer Solution, Potassium Hydrogen Phthalate
pH 4.008
Packaging Bottle
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
12930654
View Documents
Fisher Chemical
SOL 009 500
500 mL Bottle
42.00€
500mL
Please to purchase this item. Need a web account? Register with us today!
12990654
View Documents
Fisher Chemical
SOL 015 1L
1000 mL Bottle
61.00€
1000mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Buffer Solution, Potassium Hydrogen Phthalate
4.008
Bottle
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 12 months

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.