missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BTB1
Selective and ATP-competitive Kif18A inhibitor
113.00€ - 404.00€
Specifications
Purity | >99% |
---|---|
Product Type | BTB1 |
Molecular Weight (g/mol) | 297.71 |
Inhibitors | BTB1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18778198
|
Tocris Bioscience™
5399/10 |
10 mg |
113.00€
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18713439
|
Tocris Bioscience™
5399/50 |
50 mg |
404.00€
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Selective and ATP-competitive mitotic kinesin Kif18A inhibitor (IC50 = 1.7 μM); reversibly inhibits microtubule-stimulated Kif18A ATPase activity. Displays selectivity against a panel of other mitotic kinesins.Specifications
>99% | |
BTB1 | |
297.71 | |
BTB1 |
For Research Use Only