missing translation for 'onlineSavingsMsg'
Learn More

BRL 50481, Tocris Bioscience™

Product Code. 18093962
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective PDE7 inhibitor

Selective, substrate-competitive inhibitor of phosphodiesterase (PDE) 7 (Ki = 180 nM). Displays > 200-fold selectivity over PDE1B, PDE1C, PDE2, PDE3, PDE4A4 and PDE5.

Chemical Identifiers

CAS 433695-36-4
Molecular Formula C9H12N2O4S
Molecular Weight (g/mol) 244.265
InChI Key IFIUFCJFLGCQPH-UHFFFAOYSA-N
Synonym unii-03g869pr3p, benzenesulfonamide, n,n,2-trimethyl-5-nitro, 5-nitro-2,n,n-trimethylbenzenesulfonamide, d00mbg, cambridge id 6971444, n,n,2-trimethyl-5-nitro-benzenesulfonamide, brl hplc , solid, n,n,2-trimethyl-5-nitrobenzene-1-sulfonamide, 2-methyl-5-nitro n,n-dimethyl benzenesulfonamide, dimethyl 2-methyl-5-nitrophenyl sulfonyl amine
PubChem CID 2921148
IUPAC Name N,N,2-trimethyl-5-nitrobenzenesulfonamide
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])S(=O)(=O)N(C)C

Specifications

Quantity 50 mg
Formula Weight 244.26
Percent Purity >99%
Chemical Name or Material BRL 50481
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'