missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Breast Matched Tumor and Normal Tissue Lysate

Product Code. 18107087 Shop All Bio Techne Products
Click to view available options
Quantity:
2 Vials
Unit Size:
Pack of 2
This item is not returnable. View return policy

Product Code. 18107087

Brand: Novus Biologicals™ NBP247112

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Two matched 1mg/mL vials (100μg/vial) of human breast tissue lysate, one containing normal tissue and the other tumor tissue, for use in research. Lysates are guaranteed for 6 months from date of receipt.

The Breast Matched Tumor & Normal Tissue Lysate is derived from human.
TRUSTED_SUSTAINABILITY

Specifications

Tissue Breast
Quantity 2 Vials
Host Species Human
Content And Storage Store at −80°C. Avoid freeze-thaw cycles.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.