Learn More
Aalborg™ Precision Valve for Brass Flowmeter
Aalborg™ Precision Valve for Brass Flowmeter is designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
Brand:  Aalborg™ VM5-BB
 View more versions of this product
Product Code. 11758278
  • 120.00€ / Each
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For high precision
  • Designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
  • They may also be used as shut off valves.
  • Bodies of the valves are reinforced by structurally rigid metallic shells.
  • Shells are made of brass, bushings are made of plated brass or 316 stainless steel.
Specifications
Specifications
6200 mL/min.
200mL/min.
150 mm Gas/Liquid Flowmeter
Standard Valve