Learn More
Brass foil, alloy 260, 1.0mm (0.04in) thick, Thermo Scientific Chemicals
38.40€ - 70.80€
Specifications
Chemical Name or Material | Brass foil, Alloy 260 |
---|---|
Name Note | 1.0mm (0.04 in.) thick |
MDL Number | MFCD00171255 |
Solubility Information | Insoluble in water. |
Odor | Odorless |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15402217
|
Thermo Scientific Alfa Aesar
045221.GT |
150 x 150 mm |
38.40€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15412217
|
Thermo Scientific Alfa Aesar
045221.HB |
300 x 300 mm |
70.80€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Brass foil, alloy has a wide range of applications, such as appliance parts, highly formable parts, ammunition components, decorative parts of furniture and cabinets, name plates, engraving plates, overlays, legend plates, lamp fixtures, fasteners, locks, hinges, flashlight shells, connectors, fittings, circuit board relays, electronic components, switches, nuts, bolts, valve stems and valve components, radiator cores and tanks, brackets, frames, fixtures, ornamental works, and used extensively in the musical instrument, electrical equipment, plumbing, central heating, seawater lines and desalination fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Brass foil, Alloy 260 | |
MFCD00171255 | |
Odorless |
1.0mm (0.04 in.) thick | |
Insoluble in water. | |
Foil |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only