Learn More
VACUUBRAND™ MD1C + AK + EK Oil-Free Chemistry Diaphragm Vacuum Pump
An excellent choice for rotary evaporation and other laboratory applications
Brand: VACUUBRAND™ 20696620
This item is not returnable.
View return policy
Description
Suitable for benchtop rotary evaporators, and powerful enough for excellent results with high-boiling point solvents
- Extremely low maintenance, no oil changes, no oil mist
- Ultimate vacuum 1.5 Torr; free air capacity 25 liters per minute
- Corrosion resistance
- With an integrated gas ballast to provide consistent pump performance with condensed vapors
- Adds solvent recovery, including an emissions condenser; compact three-stage design is suitable for benchtop rotary evaporation and gel drying with an SDS-PAGE concentration greater than 10%
Order Info
Included solvent recovery glassware for use in evaporative applications when no cold trap is used.
Specifications
CEE plug | |
12.62 x 11.4 x 19 in. (32 x 29 x 48 cm) | |
MD1C+AK+EK | |
10.2 kg | |
60 Hz | |
48 cm | |
32 cm | |
29 cm |
230 V | |
PTFE (Pump Head) | |
22.48 lb. | |
25 L/min. | |
19 in. | |
12.62 in. | |
11.4 in. | |
230 V |