Learn More
BRAND™ Replacement Parts for Dispensette™ Organic Bottle Top Dispensers: Discharge Valve Washer
Sealing washers for BRAND Dispensette III and Dispensette Organic.
Brand: BRAND™ 6696
20.95 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
- Does not fit other dispensers
- Five each of filling washers, and discharge/SafetyPrime washers.
Specifications
Discharge Tube Seal | |
PTFE | |
Dispensette™ III, organic and digital buret III | |
Five filing valve rings and five discharge/SafetyPrime™ seals |
Promotions
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
New Offer Save Up to 92% -
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Product Content Correction
Product Title
BRAND™ Replacement Parts for Dispensette™ Organic Bottle Top Dispensers: Discharge Valve Washer > Discharge valve washer for dispensette III, organic and digital buret III