missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates cellGrade™ plus, 96-well Flat Bottom Microplates, PS

Product Code. 10793074 Shop All Brand Products
Click to view available options
Color:
Black, Clear Bottom
Clear
White, Clear Bottom, Clear Bottom
Unit Size:
Pack of 5
Pack of 50
Packaging:
1/Pk., 50/Cs.
5/Pk.
This item is not returnable. View return policy

Product Code. 10793074

Brand: BRAND™ 782031

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Sterile 96-well microplates with cellGrade™plus surface treatment for optimal cell growth. Features protein-like surface for fast cell attachment and better yield. Ideal for cultivating sensitive or fastidious cell lines, even in reduced-serum media

  • Hydrophilic surface with freely-accessible amino, carboxyl and hydroxyl groups
  • Protein-like composition allows for direct cellular attachment
  • Sterilized by B-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
  • Meet the requirements of Ph. Eur. and the USP 29
  • BIO-CERT™ CELL CULTURE QUALITY: free of pyrogens, endotoxins, human DNA, RNase, and DNase
  • Endotoxins are <0.01 EU/mL
  • Orange alphanumeric embossing for easy well identification

Order Info

Packs of 50 individually wrapped plates with lids

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Binding Property Hydrophilic
Certifications/Compliance ISO 11137, DIN EN ISO 10993
Format Standard
Lid With Lid
Array 8 x 12
Color Black, Clear Bottom
Product Type 96-well microplate
Material Polystyrene
Volume (Metric) Well 330 μL
No. of Wells 96
Sterility Sterile
Surface Treatment cellGrade Plus
Total Volume (Metric) Well 330 μL
Well Shape Flat Bottom (F-well)
For Use With (Application) Cultivation of cells in reduced serum media and fastidious cell lines
No. per Case 50/Cs.
No. per Pack 1/Pk.
Well Color Black
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.