Learn More
BRAND™ BRANDplates™ PC Membrane Single Inserts
Sterile, individual inserts
Brand:  BRAND™ 782826
318.75 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15670617
  • 348.00€ / Pack of 48
Estimated Shipment: 20-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Innovative cell culture plates and inserts for manual and automated cell and tissue culture
  • The BRANDplates™ Insert System, developed jointly with the Fraunhofer Gesellschaft, forms an innovative extension of BRAND's existing cell culture range
  • It consists of two different cell culture plates and two versions of cell culture inserts, which can also be combined with each other
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Smooth walled
    24-/6-well plates BRANDplates™
    48
    PC Membrane 1.0 μm
    13 mm