Learn More
Brady™ BradyPrinter i5100 Kit UK and EU with Workstation PWID Suite
The BradyPrinter i5100 Industrial Label Printer is smart and easy to use, with excellent performance and the ability to print more than 1200 standard identification labels, plus custom labels.
Brand:  Brady™ 198780
Product Code. 16429253
  • 2045.00€ / Each
Explore more special offers
Description
Description
Create and print professional labels with the BradyPrinter i5100 Kit. Intelligent Print (IP) technology allows the materials, printer and included Brady™ Workstation PWID Suite software to communicate.
  • Change printer settings easily with intuitive touchscreen
  • Change out materials quickly with specially designed materials that easily lock into place
  • Connect easily with a variety of connection ports
  • Keep printer running smoothly with interchangable rollers
  • Prints more than 1200 standard identificaiton labels plus custom labels
  • Includes Brady™ Workstation Product and Wire Identification Software Suite
Specifications
Specifications
Color LCD Touchscreen
9.5 in.
12.5 in.
17 in.
Not Autoclavable
Monochrome
Arabic, Bulgarian, Chinese (Tradditional), Chinese (Simplified), Croatian, Czech, Danish, Dutch, English, Finnish, French, German, Greek, Hungarian, Italian, Lithuanian, Norwegian, Polish, Portuguese, Romanian, Russian, Serbian, Slovenian, Spanish, Swedish, Turkish
i5100
600 dpi
Windows™ 10, Windows™ 7, Windows™ 8, Windows™ Servers 2012, 2012 R2, and 2016
14 lb.
Asset Tracking, Circuit Board Labeling, General Identification
110/240 V
Thermal Transfer
241 mm
318 mm
437 mm
Non-sterile
i5100 Printer, Power Cord, USB Cable, Product CD with Driver, User Manual, RFID Enabled Roll Holder, Ribbon Take Up Spindle, Brady™ Workstation PWID Suite
256 MB RAM, 50 MB IFFS Flash, SD Card, Slot Up To 512 GB
150 mm/s
Bluetooth™, Ethernet, Serial, USB
0°C to 40°C
6.2 kg
2 Year