Learn More
Brady™ BradyJet J2000 Colour Label Printer - EU with Brady Workstation LAB Suite
The BradyJet J2000 Colour Label Printer is a practical and easy to use, full colour inkjet label printer that can print reliable, high-resolution signs, facility and laboratory sample labels.
Brand:  Brady™ 199966
1798.49 EUR valid until 2024-12-31
Use promo code "21918" to get your promotional price.
Product Code. 16140501
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
The perfect combo. Get the impact of colour—that’s wildly durable—to create a safer, more efficient workplace. Rely on the BradyJet J2000 Colour Label Printer when durable colour matters most.
  • Works great in a multitude of industries, from lab to safety and more
  • Compact design means it can snuggle in to tight spaces
  • On-demand, mid-volume inkjet solution creates photo-quality colour labels in a single pass
  • More colour options than you’d ever need—16 million!
  • Custom identification without the high cost of outsourced printing
  • Includes Brady Workstation Lab Suite to your solution and make label creation even easier
  • Setup is stress free – Contact our Brady Technical Support Team and we will schedule a free phone setup appointment
Specifications
Specifications
None
389 x 264.00 mm
This kit contains a J2000 printer, a power cord and AC adapter, a USB cable, a Quick Start Guide, an ink cartridge, a material roll for printhead alignment, a USB stick with drivers and manuals and a Brady Workstation Laboratory Suite
4800 dpi
Inkjet
Multi-color
63.50 mm/sec.
100/240 V