Learn More
Invitrogen™ Bovine Rhodopsin (aa 338-348) Synthetic Peptide
Synthetic Peptide
Brand:  Invitrogen™ PEP174
Product Code. 11544830
  • 191.00€ / 50µg
Explore more special offers
Description
Description
Bovine Rhodopsin (residues 338-348) Synthetic Peptide, PEP-174, from Invitrogen.
PEP-174 is a 12 amino acid synthetic peptide whose sequence is derived from bovine rhodopsin protein. The sequence of this peptide is (amino to carboxy terminus): C-S (338)-K-T-E-T-S-Q-V-A-P-A (348)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and bovine rhodopsin, catalog # PA1-729. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
509933
purified
Bovine Rhodopsin (aa 338-348)
-20°C. Avoid freeze-thaw cycles.
Lyophilized
Control
Yes
Peptide
RUO - research use only
Bovine
P02699
0.5 mg/mL
Synthetic Peptide
peptide with no preservative
RHO
No preservative
50 μg
C-S(338)-K-T-E-T-S-Q-V-A-P-A(348)