Learn More
Fisherbrand™ Borosilicate Glass Medium Wall Rimless Test Tube
Manufactured from borosilicate glass with a medium wall design. Fisherbrand™ Borosilicate Glass Medium Wall Rimless Test Tubes are ideal for chemical and biological work.
Brand:  Fisherbrand™ FB63145
 View more versions of this product
Product Code. 11952208
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Features: rimless medium walls
  • Grade: 3.3 borosilicate glass test tubes
  • Complies to: ISO 4142
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 4142
Borosilicate Glass
No
125 mm
100 Pack
16 mm