Learn More
Rogo Sampaic™ Glass Measuring Cup
Brand:  Rogo Sampaic™ CBD021
 View more versions of this product
Product Code. 11711816
  • 495.00€ / Pack of 10
Estimated Shipment: 10-04-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Borosilicate Glass
100 mL
140 mm