Learn More
Fisherbrand™ Borosilicate Glass Heavy Wall Rimmed Test Tubes
Product Code. p-8002023 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric):
16 mm
24 mm
Length (Metric):
125 mm
150 mm
Unit Size:
Pack of 100
Pack of 50
Product Code. 11982208
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Manufactured from borosilicate glass with a heavy wall design for strong resistance to heat and chemicals. Fisherbrand™ Borosilicate Glass Heavy Wall Rimmed Test Tubes are ideal for chemical and biological work.
  • Features: rimmed heavy walls, superior mechanical strength
  • Grade: 3.3 borosilicate glass test tubes
  • Complies to: ISO 4142
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 4142
Length (Metric) 125 mm
Material Borosilicate Glass
Quantity 100 Pack
Rimmed Yes
Diameter (Metric) 16 mm