Learn More
Fisherbrand™ Borosilicate Glass Graduated Cylinder
Product Code. p-8002018 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Graduations:
0.1 mL
0.2 mL
0.5 mL
1 mL
2 mL
5 mL
10 mL
20 mL
Unit Size:
Each
Pack of 2
Product Code. 11577702
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Measure liquids with blue ceramic scale graduated glass cylindars. The Fisherbrand™ borosilicate glass graduated cylindawith hexagonal base and spout
  • DIN 12680, BS 604, ISO 4788
  • Graduated in blue ceramic
  • Borosilicate Glass
  • Hexagonal base
  • Spouted
  • Individual reorder code on each product
TRUSTED_SUSTAINABILITY
Specifications
Base Type Hexagonal
Scale Color Blue
Type Graduated Cylinder
Material Borosilicate Glass
Certifications/Compliance BS 604, DIN 12680, ISO 4788
Class B
Capacity (Metric) 1000 mL
Scale Blue Ceramic
Graduations 10 mL