Learn More
Rogo Sampaic™ Glass Coverslips
Preserve and protect specimens on slides.
Brand:  Rogo Sampaic™ BPD030
 View more versions of this product
Product Code. 13286718
  • 90.00€ / Pack of 100
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Available in a range of sizes
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
Coverslip
Soda Lime Glass
100/Pk.
Rectangle and Square
14 mm