Learn More
Rogo Sampaic™ Glass Coverslips
Preserve and protect specimens on slides.
Brand:  Rogo Sampaic™ BPD029
 View more versions of this product
Product Code. 13286708
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Available in a range of sizes
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
Coverslip
Borosilicate Glass
100/Pk.
Rectangle and Square
22 x 32 mm